3-(Methylthio)benzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H27200-03 H27200-14 H27200-06
AAH27200-03EA 49.3 USD
AAH27200-03 AAH27200-14 AAH27200-06
3-(Methylthio)benzoic acid 97%
3-(Methylthio)benzoic acid

Purity: 97% CAS No.: 825-99-0 Molecular Formula: C8H8O2S Formula Weight: 168.21 Melting Point: 126-132° MDL No.: MFCD00156993

Formula: C₈H₈O₂S
Melting Pt: 126…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00156993
CAS Number: 825-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR