D(-)-allo-Isoleucine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
H-D-Allo-Ile-OH, (2R,3S)-(-)-allo-Isoleucine, H-D-allo-Ile-OH
H27189-06
H27189-MD
AAH27189-03EA
269.72
USD
AAH27189-03
AAH27189-06
AAH27189-MD
D(-)-allo-Isoleucine 97%
D(-)-allo-Isoleucine
Purity: 97%
CAS No.: 1509-35-9
Molecular Formula: C6H13NO2
Formula Weight: 131.18
Melting Point: ca 285° dec.
Beilstein Reference: 1721794
Formula:
C₆H₁₃NO₂ MW: 131.17 g/mol Melting Pt: ∼ 285 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00066445 CAS Number: 1509-35-9 EINECS: 216-143-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...