1-Hexyl-3-methylimidazolium chloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-n-Hexyl-3-methylimidazolium chloride
H27178-06
H27178-18
AAH27178-06EA
39
USD
AAH27178-06
AAH27178-18
1-Hexyl-3-methylimidazolium chloride 98%
1-Hexyl-3-methylimidazolium chloride
Purity: 98%
CAS No.: 171058-17-6
Molecular Formula: C10H19N2Cl
Formula Weight: 202.72
Refractive Index: 1.5165
Formula:
C₁₀H₁₉ClN₂ MW: 202.73 Melting Pt: -85 Density: 1.05 Storage Temperature: Ambient |
MDL Number:
MFCD03093289 CAS Number: 171058-17-6 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...