Methyldiphenylphosphine 99%

Supplier: Thermo Scientific Chemicals
Warning

H27159-03 H27159-18 H27159-09
AAH27159-03EA 31.3 USD
AAH27159-03 AAH27159-18 AAH27159-09
Methyldiphenylphosphine 99%
Methyldiphenylphosphine

Purity: 99% CAS No.: 1486-28-8 Molecular Formula: (C6H5)2PCH3 Formula Weight: 200.21 Flash Point: 110°(230°F) Boiling Point: 160°/15mm MDL No.: MFCD00008508

Formula: C₁₃H₁₃P
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 28 °C
Density: 1.0784 g/cm³ (15 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008508
CAS Number: 1486-28-8
EINECS: 216-065-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR