1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate 99%
Supplier: Thermo Scientific Chemicals
H27141-06
H27141-18
AAH27141-06EA
56.5
USD
AAH27141-06
AAH27141-18
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate 99%
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate
Purity: 99%
CAS No.: 402846-78-0
Molecular Formula: C9H17F4N2B
Formula Weight: 240.05
MDL No.: MFCD03427618
Formula:
C₉H₁₇BF₄N₂ Density: 1,198 Storage Temperature: Ambient |
MDL Number:
MFCD03427618 CAS Number: 402846-78-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...