4-Methoxy-o-toluic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H27096-06
H27096-03
AAH27096-06EA
114
USD
AAH27096-06
AAH27096-03
4-Methoxy-o-toluic acid 97%
4-Methoxy-o-toluic acid
Purity: 97%
CAS No.: 6245-57-4
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 177-181°
MDL No.: MFCD00020291
Formula:
C₉H₁₀O₃ MW: 166.18 Melting Pt: 177…181 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020291 CAS Number: 6245-57-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...