5-Bromo-o-cresol 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Bromo-3-hydroxy-4-methylbenzene, 5-Bromo-2-methylphenol
H26956-03
H26956-09
AAH26956-03EA
35.4
USD
AAH26956-03
AAH26956-09
5-Bromo-o-cresol 95%
5-Bromo-o-cresol
Purity: 95%
CAS No.: 36138-76-8
Molecular Formula: C7H7BrO
Formula Weight: 187.04
Melting Point: 77-80°
MDL No.: MFCD08061906
Formula:
C₇H₇BrO MW: 187.04 g/mol Melting Pt: 77…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD08061906 CAS Number: 36138-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...