5-Bromo-3-nitrosalicylaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26927-06
H26927-03
AAH26927-06EA
138
USD
AAH26927-06
AAH26927-03
5-Bromo-3-nitrosalicylaldehyde 97%
5-Bromo-3-nitrosalicylaldehyde
Purity: 97%
CAS No.: 16634-88-1
Molecular Formula: C7H4BrNO4
Formula Weight: 246.02
Melting Point: 125-128°
MDL No.: MFCD00130110
Formula:
C₇H₄BrNO₄ Melting Pt: 125…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00130110 CAS Number: 16634-88-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...