2,5-Dibromothiazole 97%

Supplier: Thermo Scientific Chemicals
Warning

H26909-06 H26909-14
AAH26909-06EA 128 USD
AAH26909-06 AAH26909-14
2,5-Dibromothiazole 97%
2,5-Dibromothiazole

Purity: 97% CAS No.: 4175-78-4 Molecular Formula: C3HBr2NS Formula Weight: 242.92 Melting Point: 47-49° MDL No.: MFCD00016891

Formula: C₃HBr₂NS
Melting Pt: 47…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00016891
CAS Number: 4175-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR