1-Ethyl-3-methylimidazolium dicyanamide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Basionics VS 03, EMIM DCA

H26901-06 H26901-18
AAH26901-06EA 117 USD
AAH26901-06 AAH26901-18
1-Ethyl-3-methylimidazolium dicyanamide 98%
1-Ethyl-3-methylimidazolium dicyanamide

Purity: 98% CAS No.: 370865-89-7 Molecular Formula: C8H11N5 Formula Weight: 177.21 MDL No.: MFCD08276373

Formula: C₈H₁₁N₅
MW: 177.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08276373
CAS Number: 370865-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR