4-(Methylthio)phenylacetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26888-03
H26888-06
AAH26888-03EA
27.1
USD
AAH26888-03
AAH26888-06
4-(Methylthio)phenylacetic acid 97%
4-(Methylthio)phenylacetic acid
Purity: 97%
CAS No.: 16188-55-9
Molecular Formula: C9H10O2S
Formula Weight: 182.24
Melting Point: 98-102°
MDL No.: MFCD00192325
Formula:
C₉H₁₀O₂S MW: 182.24 Melting Pt: 98…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00192325 CAS Number: 16188-55-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...