7-Fluoroisatin 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 7-Fluoro-1H-indole-2,3-dione, 7-Fluoroindoline-2,3-dione

H26827-03 H26827-06 H26827-14
AAH26827-03EA 30.2 USD
AAH26827-03 AAH26827-06 AAH26827-14
7-Fluoroisatin 97%
7-Fluoroisatin

Purity: 97% CAS No.: 317-20-4 Molecular Formula: C8H4FNO2 Formula Weight: 165.12 Melting Point: 193-195° MDL No.: MFCD01569508

Formula: C₈H₄FNO₂
MW: 165.12 g/mol
Melting Pt: 193…195 °C
Storage Temperature: Ambient
MDL Number: MFCD01569508
CAS Number: 317-20-4
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR