4-Heptylaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

H26793-06 H26793-14
AAH26793-03EA 18.9 USD
AAH26793-03 AAH26793-06 AAH26793-14
4-Heptylaniline 98%
4-Heptylaniline

Purity: 98% CAS No.: 37529-27-4 Molecular Formula: C13H21N Formula Weight: 191.31 Flash Point: 110°(230°F) Refractive Index: 1.5200 Boiling Point: 159°/18mm MDL No.: MFCD00007928

Formula: C₁₃H₂₁N
MW: 191.32 g/mol
Boiling Pt: 159 °C/18 mm
Density: 0.91
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007928
CAS Number: 37529-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR