Yeast Extract, Thermo Scientific
Supplier: Thermo Scientific Chemicals
H26769-36
H26769-22
AAH26769-36EA
183.14
USD
AAH26769-36
AAH26769-22
Yeast Extract, Thermo Scientific
Media for Microbiology
Dehydrated Media for Microbiology
CAS No.: 8013-01-2
MDL No.: MFCD00132599
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...