Peptone from Meat, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Peptone is used in nutrient media for growing bacteria and fungiMeat Peptone is used for routine and mass scale cultivation of organisms, which is utilized for the production of antibiotics, enzymes and vitamin production. It is also useful for the cultivation of a variety of microorganisms. Further, it is used in culture media, industry fermentation and the pharmaceutical industry.
- Soluble in water. Insoluble in alcohol.
- Hygroscopic. Incompatible with strong oxidizing agents.
CAS No.: 73049-73-7
MDL No.: MFCD00131829
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...