2-Fluoro-p-toluic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H26688-03 H26688-06
AAH26688-03EA 38.1 USD
AAH26688-03 AAH26688-06
2-Fluoro-p-toluic acid 97%
2-Fluoro-p-toluic acid

Purity: 97% CAS No.: 7697-23-6 Molecular Formula: C8H7FO2 Formula Weight: 154.14 Melting Point: 186-189° MDL No.: MFCD03092906

Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 186…189 °C
Storage Temperature: Ambient
MDL Number: MFCD03092906
CAS Number: 7697-23-6
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR