4-Pentylaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amylaniline, 4-n-Pentylaniline, 4-Pentylaniline

H26569-06 H26569-14
AAH26569-06EA 53.8 USD
AAH26569-06 AAH26569-14
4-Pentylaniline 98%
4-Pentylaniline

Purity: 98% CAS No.: 33228-44-3 Molecular Formula: C11H17N Formula Weight: 163.26 Flash Point: 110°(230°F) Refractive Index: 1.53 Boiling Point: 130° MDL No.: MFCD00007926

Formula: C₁₁H₁₇N
MW: 163.26 g/mol
Boiling Pt: 130 °C
Density: 0.919
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007926
CAS Number: 33228-44-3
EINECS: 251-409-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR