2-Fluoro-6-methoxyphenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26471-03
H26471-06
AAH26471-03EA
71.5
USD
AAH26471-03
AAH26471-06
2-Fluoro-6-methoxyphenol 97%
2-Fluoro-6-methoxyphenol
Purity: 97%
CAS No.: 73943-41-6
Molecular Formula: C7H7FO2
Formula Weight: 142.13
Flash Point: 103°(218°F)
Refractive Index: 1.52
Boiling Point: 130-131°/36mm
MDL No.: MFCD00075140
Formula:
C₇H₇FO₂ MW: 142.13 g/mol Boiling Pt: 130…131 °C/36 mm Density: 1.23 Flash Pt: 103 °C(218 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00075140 CAS Number: 73943-41-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...