2-Bromo-5-(trifluoromethoxy)phenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26389-06
H26389-14
AAH26389-06EA
110
USD
AAH26389-06
AAH26389-14
2-Bromo-5-(trifluoromethoxy)phenol 97%
2-Bromo-5-(trifluoromethoxy)phenol
Purity: 97%
CAS No.: 205371-26-2
Molecular Formula: C7H4BrF3O2
Formula Weight: 257.01
Refractive Index: 1.4815
Formula:
C₇H₄BrF₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD06660162 CAS Number: 205371-26-2 EINECS: None |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...