4'-Chloro-3'-fluoroacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(4-Chloro-3-fluorophenyl)ethanone

H26373-03 H26373-06 H26373-14
AAH26373-03EA 83.5 USD
AAH26373-03 AAH26373-06 AAH26373-14
4'-Chloro-3'-fluoroacetophenone 97%
4'-Chloro-3'-fluoroacetophenone

Purity: 97% CAS No.: 151945-84-5 Molecular Formula: C8H6ClFO Formula Weight: 172.59 MDL No.: MFCD04115858

Formula: C₈H₆ClFO
MW: 172.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115858
CAS Number: 151945-84-5
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR