2-Bromo-5-fluoroanisole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Bromo-4-fluoro-2-methoxybenzene
H26325-06
H26325-14
AAH26325-06EA
135
USD
AAH26325-06
AAH26325-14
2-Bromo-5-fluoroanisole 97%
2-Bromo-5-fluoroanisole
Purity: 97%
CAS No.: 450-88-4
Molecular Formula: C7H6BrFO
Formula Weight: 205.03
Flash Point: 90°(194°F)
Refractive Index: 1.5460
Boiling Point: 143-145°
MDL No.: MFCD04973752
Formula:
C₇H₆BrFO MW: 205.03 g/mol Boiling Pt: 143…145 °C Density: 1.599 Flash Pt: 90 °C(194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD04973752 CAS Number: 450-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...