4-Hexylaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26297-14
H26297-06
AAH26297-14EA
171
USD
AAH26297-14
AAH26297-06
4-Hexylaniline 98%
4-Hexylaniline
Purity: 98%
CAS No.: 33228-45-4
Molecular Formula: C12H19N
Formula Weight: 177.29
Flash Point:
Formula:
C₁₂H₁₉N MW: 177.29 g/mol Boiling Pt: 283…285 °C Density: 0.91 g/cm³ (20 °C) Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007927 CAS Number: 33228-45-4 EINECS: 251-409-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...