4-Ethynylanisole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(4-Methoxyphenyl)ethyne, (4-Methoxyphenyl)acetylene, 1-Ethynyl-4-methoxybenzene, 1-Methoxy-4-ethynylbenzene
H26283-03
H26283-06
AAH26283-03EA
77.7
USD
AAH26283-03
AAH26283-06
4-Ethynylanisole 99%
4-Ethynylanisole
Purity: 99%
CAS No.: 768-60-5
Molecular Formula: C9H8O
Formula Weight: 132.16
Flash Point: 82°(180°F)
Refractive Index: 1.563
Melting Point: 27-29°
Boiling Point: 87-91°/11mm
MDL No.: MFCD00168815
Formula:
C₉H₈O MW: 132.16 g/mol Boiling Pt: 87…91 °C/11 mm Melting Pt: 27…29 °C Density: 1.019 Flash Pt: 82 °C(180 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00168815 CAS Number: 768-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...