2-Iodopyrazine 95%

Supplier: Thermo Scientific Chemicals
Warning

H26244-06 H26244-03
AAH26244-06EA 150 USD
AAH26244-06 AAH26244-03
2-Iodopyrazine 95%
2-Iodopyrazine

Purity: 95% CAS No.: 32111-21-0 Molecular Formula: C4H3IN2 Formula Weight: 205.98 Flash Point: 110°(230°F) Refractive Index: 1.655 Boiling Point: 109-110°/34mm MDL No.: MFCD01319019

Formula: C₄H₃IN₂
MW: 205.99 g/mol
Boiling Pt: 109…110 °C/34 mm
Density: 2.086
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01319019
CAS Number: 32111-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR