3-Bromo-2,6-difluorobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

H26213-06 H26213-14
AAH26213-06EA 148 USD
AAH26213-06 AAH26213-14
3-Bromo-2,6-difluorobenzaldehyde 98%
3-Bromo-2,6-difluorobenzaldehyde

Purity: 98% CAS No.: 398456-82-1 Molecular Formula: C7H3BrF2O Formula Weight: 221.00 Melting Point: 53-57° MDL No.: MFCD08282773

Formula: C₇H₃BrF₂O
Melting Pt: 53…57 °C
Storage Temperature: Ambient
MDL Number: MFCD08282773
CAS Number: 398456-82-1
EINECS: None

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR