4-Bromo-2,5-difluorophenol 99%

Supplier: Thermo Scientific Chemicals
Warning

H26212-14 H26212-06
AAH26212-14EA 260 USD
AAH26212-14 AAH26212-06
4-Bromo-2,5-difluorophenol 99%
4-Bromo-2,5-difluorophenol

Purity: 99% CAS No.: 486424-36-6 Molecular Formula: C6H3BrF2O Formula Weight: 208.99 Melting Point: 51-53° MDL No.: MFCD04973751

Formula: C₆H₃BrF₂O
Melting Pt: 51…53 °C
Storage Temperature: Ambient
MDL Number: MFCD04973751
CAS Number: 486424-36-6
EINECS: None

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR