2-Bromo-3,5-difluorophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

H26203-06 H26203-03
AAH26203-06EA 172 USD
AAH26203-06 AAH26203-03
2-Bromo-3,5-difluorophenol 98%
2-Bromo-3,5-difluorophenol

Purity: 98% CAS No.: 325486-43-9 Molecular Formula: C6H3BrF2O Formula Weight: 208.99 Refractive Index: 1.529

Formula: C₆H₃BrF₂O
MW: 208.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861127
CAS Number: 325486-43-9
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR