3-Bromo-5-(trifluoromethyl)benzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Bromo-5-trifluoromethylbenzaldehyde

H26197-06 H26197-03
AAH26197-06EA 103 USD
AAH26197-06 AAH26197-03
3-Bromo-5-(trifluoromethyl)benzaldehyde 97%
3-Bromo-5-(trifluoromethyl)benzaldehyde

Purity: 97% CAS No.: 477535-41-4 Molecular Formula: C8H4BrF3O Formula Weight: 253.02 Refractive Index: 1.5110

Formula: C₈H₄BrF₃O
MW: 253.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08059506
CAS Number: 477535-41-4
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR