(2,4,5-Trifluorophenyl)acetic acid 94%

Supplier: Thermo Scientific Chemicals
Warning

H26190-03 H26190-06
AAH26190-03EA 30.3 USD
AAH26190-03 AAH26190-06
(2,4,5-Trifluorophenyl)acetic acid 94%
(2,4,5-Trifluorophenyl)acetic acid

Purity: 94% CAS No.: 209995-38-0 Molecular Formula: C8H5F3O2 Formula Weight: 190.12 Melting Point: 121-124° MDL No.: MFCD00082479

Formula: (C₆H₂F₃)CH₂COOH
Melting Pt: 121…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00082479
CAS Number: 209995-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR