3,5-Difluorophenylhydrazine hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (3,5-Difluorophenyl)hydrazine hydrochloride

H26189-06 H26189-03
AAH26189-06EA 127 USD
AAH26189-06 AAH26189-03
3,5-Difluorophenylhydrazine hydrochloride 97%
3,5-Difluorophenylhydrazine hydrochloride

Purity: 97% CAS No.: 134993-88-7 Molecular Formula: C6H6F2N2.HCl Formula Weight: 180.59 Melting Point: 264-266° MDL No.: MFCD03094171

Formula: C₆H₇Cl₃N₂
MW: 213.49 g/mol
Melting Pt: 264…266 °C
Storage Temperature: Ambient
MDL Number: MFCD03094171
CAS Number: 502496-27-7
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR