4-Fluoro-2-methoxybenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

H26179-06 H26179-03
AAH26179-06EA 88 USD
AAH26179-06 AAH26179-03
4-Fluoro-2-methoxybenzoic acid 99%
4-Fluoro-2-methoxybenzoic acid

Purity: 99% CAS No.: 395-82-4 Molecular Formula: C8H7FO3 Formula Weight: 170.14 Melting Point: 134-136° MDL No.: MFCD00673010

Formula: C₈H₇FO₃
Melting Pt: 134…136 °C
Storage Temperature: Ambient
MDL Number: MFCD00673010
CAS Number: 395-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR