2-Fluoro-5-iodobenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H26175-03 H26175-06
AAH26175-03EA 32.2 USD
AAH26175-03 AAH26175-06
2-Fluoro-5-iodobenzoic acid 97%
2-Fluoro-5-iodobenzoic acid

Purity: 97% CAS No.: 124700-41-0 Molecular Formula: C7H4FIO2 Formula Weight: 266.01 Melting Point: 162-165° MDL No.: MFCD03094517

Formula: C₇H₄FIO₂
MW: 266.01 g/mol
Melting Pt: 162…165 °C
Storage Temperature: Ambient
MDL Number: MFCD03094517
CAS Number: 124700-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR