5-Amino-α,α,α-trifluoro-m-toluic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H26169-06
H26169-03
AAH26169-06EA
336
USD
AAH26169-06
AAH26169-03
5-Amino-α,α,α-trifluoro-m-toluic acid 97%
5-Amino-α,α,α-trifluoro-m-toluic acid
Purity: 97%
CAS No.: 328-68-7
Molecular Formula: C8H6F3NO2
Formula Weight: 205.14
Melting Point: 142-146°
MDL No.: MFCD00236641
Formula:
C₈H₆F₃NO₂ Melting Pt: 142…146 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236641 CAS Number: 328-68-7 EINECS: None |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...