2,6-Difluoro-4-hydroxybenzoic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

H26134-03 H26134-MD
AAH26134-03EA 126 USD
AAH26134-03 AAH26134-MD
2,6-Difluoro-4-hydroxybenzoic acid 95%
2,6-Difluoro-4-hydroxybenzoic acid

Purity: 95% CAS No.: 214917-68-7 Molecular Formula: C7H4F2O3 Formula Weight: 174.10 Melting Point: 170-171° dec. MDL No.: MFCD03094500

Formula: C₇H₄F₂O₃
Melting Pt: 170…171 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03094500
CAS Number: 214917-68-7
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR