2-Amino-5-phenylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Phenylpyridin-2-amine, 5-Phenyl-2-pyridylamine

H26002-03 H26002-MD
AAH26002-03EA 162 USD
AAH26002-03 AAH26002-MD
2-Amino-5-phenylpyridine 97%
2-Amino-5-phenylpyridine

Purity: 97% CAS No.: 33421-40-8 Molecular Formula: C11H10N2 Formula Weight: 170.21 Melting Point: 133-137° MDL No.: MFCD01692452

Formula: C₁₁H₁₀N₂
MW: 170.21 g/mol
Melting Pt: 133…137 °C
Storage Temperature: Ambient
MDL Number: MFCD01692452
CAS Number: 33421-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR