2-Bromo-4-chloro-1-fluorobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Bromo-4-fluorochlorobenzene, 2-Bromo-4-chlorofluorobenzene, 1-Bromo-3-chloro-6-fluorobenzene, 3-Bromo-1-chloro-4-fluorobenzene

H25935-06 H25935-14
AAH25935-06EA 78.1 USD
AAH25935-06 AAH25935-14
2-Bromo-4-chloro-1-fluorobenzene 97%
2-Bromo-4-chloro-1-fluorobenzene

Purity: 97% CAS No.: 1996-30-1 Molecular Formula: C6H3BrClF Formula Weight: 209.45 Flash Point: 85°(185°F) Refractive Index: 1.554 Boiling Point: 180° MDL No.: MFCD00672934

Formula: C₆H₃BrClF
MW: 209.44 g/mol
Boiling Pt: 180 °C
Density: 1.74
Flash Pt: 85 °C(185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00672934
CAS Number: 1996-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR