2,7-Dibromo-9-fluorenone 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,7-Dibromo-9H-fluoren-9-one
H25919-03
H25919-06
AAH25919-03EA
33.7
USD
AAH25919-03
AAH25919-06
2,7-Dibromo-9-fluorenone 96%
2,7-Dibromo-9-fluorenone
Purity: 96%
CAS No.: 14348-75-5
Molecular Formula: C13H6Br2O
Formula Weight: 337.99
Melting Point: 195-196°
MDL No.: MFCD00010790
Formula:
C₁₃H₆Br₂O MW: 338 g/mol Melting Pt: 195…196 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010790 CAS Number: 14348-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...