2-Bromo-3-(trifluoromethyl)pyridine 98%

Supplier: Thermo Scientific Chemicals
Danger

H25822-03 H25822-06
AAH25822-03EA 80 USD
AAH25822-03 AAH25822-06
2-Bromo-3-(trifluoromethyl)pyridine 98%
2-Bromo-3-(trifluoromethyl)pyridine

Purity: 98% CAS No.: 175205-82-0 Molecular Formula: C6H3BrF3N Formula Weight: 226.00 Flash Point: 29°(84°F) Melting Point: 37-39° Boiling Point: 76°/8mm MDL No.: MFCD00153084

Formula: C₆H₃BrF₃N
MW: 226 g/mol
Boiling Pt: 76 °C/8 mm
Melting Pt: 37…39 °C
Flash Pt: 29 °C (84 °F)
Storage Temperature: Ambient
MDL Number: MFCD00153084
CAS Number: 175205-82-0
UN: 2926
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR