(1-Butyl)triphenylphosphonium chloride

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Butyltriphenylphosphonium chloride

H25796-14 H25796-22
AAH25796-14EA 53.6 USD
AAH25796-14 AAH25796-22
(1-Butyl)triphenylphosphonium chloride
(1-Butyl)triphenylphosphonium chloride

CAS No.: 13371-17-0 Molecular Formula: C22H24ClP Formula Weight: 354.86 Melting Point: 225-228° UN No.: 3464 MDL No.: MFCD00040546

Formula: H₃C(CH₂)₃P(C₆H₅)₃Cl
MW: 354.86 g/mol
Melting Pt: 222 - 226 °C
Flash Pt: ≥ 270 °C
Storage Temperature: Ambient
MDL Number: MFCD00040546
CAS Number: 13371-17-0
EINECS: 236-443-3
UN: 3464
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR