ω-Bromo-2-hydroxyacetophenone 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Hydroxyphenacyl bromide, 2-Bromo-1-(2-hydroxyphenyl)ethan-1-one, 2-Bromo-1-(2-hydroxyphenyl)ethanone, α-Bromo-2-hydroxyacetophenone, 2-Bromo-2'-hydroxyacetophenone

B25722-06 B25722-14
AAB25722-06EA 150 USD
AAB25722-06 AAB25722-14
ω-Bromo-2-hydroxyacetophenone 97%
ω-Bromo-2-hydroxyacetophenone

Purity: 97% CAS No.: 2491-36-3 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Melting Point: 45-47° UN No.: 3263 MDL No.: MFCD01727570

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Melting Pt: 45…47 °C
Storage Temperature: Ambient
MDL Number: MFCD01727570
CAS Number: 2491-36-3
UN: 3263
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR