5-Bromo-1-indanone 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Bromo-2,3-dihydro-1H-inden-1-one
B25707-03
B25707-06
AAAB25707-03EA
40.3
USD
AAAB25707-03
AAAB25707-06
5-Bromo-1-indanone 97+%
5-Bromo-1-indanone
Purity: 97+%
CAS No.: 34598-49-7
Molecular Formula: C9H7BrO
Formula Weight: 211.06
Melting Point: 125-129°
Beilstein Reference: 2357199
Formula:
C₉H₇BrO MW: 211.06 g/mol Boiling Pt: 280 °C (1013 hPa) Melting Pt: 125…129 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082718 CAS Number: 34598-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...