2-Amino-4-chlorothiophenol 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Amino-4-chlorobenzenethiol, 2-Amino-4-chlorophenyl mercaptan, 5-Chloro-2-mercaptoaniline

B25640-06 B25640-14
AAAB25640-06EA 75.3 USD
AAAB25640-06 AAAB25640-14
2-Amino-4-chlorothiophenol 96%
2-Amino-4-chlorothiophenol

Purity: 96% CAS No.: 1004-00-8 Molecular Formula: C6H6ClNS Formula Weight: 159.64 Odor: Unpleasant Melting Point: 52-59° UN No.: 1759 MDL No.: MFCD00792528

Formula: C₆H₆ClNS
MW: 159.64 g/mol
Melting Pt: 52…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00792528
CAS Number: 1004-00-8
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR