1-(4-Methoxyphenyl)imidazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(4-Methoxyphenyl)-1H-imidazole

B25621-03 B25621-06
AAB25621-03EA 41.4 USD
AAB25621-03 AAB25621-06
1-(4-Methoxyphenyl)imidazole 98%
1-(4-Methoxyphenyl)imidazole

Purity: 98% CAS No.: 10040-95-6 Molecular Formula: C10H10N2O Formula Weight: 174.20 Odor: Odorless Melting Point: 64-67° Boiling Point: 120°/0.1mm MDL No.: MFCD00060492

Formula: C₁₀H₁₀N₂O
MW: 174.2 g/mol
Boiling Pt: 120 °C/0.1 mm
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00060492
CAS Number: 10040-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR