2-Bromo-6-methoxyphenol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25535-03
B25535-14
B25535-06
AAB25535-03EA
59.1
USD
AAB25535-03
AAB25535-14
AAB25535-06
2-Bromo-6-methoxyphenol 98+%
2-Bromo-6-methoxyphenol
Purity: 98+%
CAS No.: 28165-49-3
Molecular Formula: C7H7BrO2
Formula Weight: 203.04
Melting Point: 62-65°
MDL No.: MFCD08146628
Formula:
C₇H₇BrO₂ Melting Pt: 62…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD08146628 CAS Number: 28165-49-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...