4-Nitro-o-anisic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25523-03
B25523-06
AAAB25523-03EA
21.3
USD
AAAB25523-03
AAAB25523-06
4-Nitro-o-anisic acid 98%
4-Nitro-o-anisic acid
Purity: 98%
CAS No.: 2597-56-0
Molecular Formula: C8H7NO5
Formula Weight: 197.15
Melting Point: 145-148°
MDL No.: MFCD00216573
Formula:
C₈H₇NO₅ Melting Pt: 145…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216573 CAS Number: 2597-56-0 EINECS: 219-998-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...