3-Benzoylbenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25519-06
B25519-14
AAAB25519-06EA
63.09
USD
AAAB25519-06
AAAB25519-14
3-Benzoylbenzoic acid 98%
3-Benzoylbenzoic acid
Purity: 98%
CAS No.: 579-18-0
Molecular Formula: C14H10O3
Formula Weight: 226.23
Odor: Odorless
Melting Point: 162-166°
Beilstein Reference: 2212366
MDL No.: MFCD00002518
Formula:
C₁₄H₁₀O₃ Boiling Pt: 429 °C (1013 hPa) Melting Pt: 162 - 166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002518 CAS Number: 579-18-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...