2-Bromo-5-chloroanisole 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Bromo-4-chloro-2-methoxybenzene

B25512-14 B25512-06
AAAB25512-14EA 513 USD
AAAB25512-14 AAAB25512-06
2-Bromo-5-chloroanisole 98+%
2-Bromo-5-chloroanisole

Purity: 98+% CAS No.: 174913-09-8 Molecular Formula: C7H6BrClO Formula Weight: 221.48 Refractive Index: 1.5875

Formula: C₇H₆BrClO
MW: 221.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03790889
CAS Number: 174913-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR