3-Aminothiophenol 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Aminophenyl mercaptan, 3-Mercaptoaniline, 3-Aminobenzenethiol

B25473-06 B25473-14
AAAB25473-06EA 69.1 USD
AAAB25473-06 AAAB25473-14 AAAB25473-22
3-Aminothiophenol 97%
3-Aminothiophenol

Purity: 97% CAS No.: 22948-02-3 Molecular Formula: C6H7NS Formula Weight: 125.19 Flash Point: 100°(212°F) Refractive Index: 1.6590 Boiling Point: 123-125°/8mm UN No.: 3267 MDL No.: MFCD00007791

Formula: C₆H₇NS
MW: 125.19 g/mol
Boiling Pt: 123…125 °C/8 mm
Density: 1.1859 g/cm³ (20 °C)
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007791
CAS Number: 22948-02-3
EINECS: 245-344-4
UN: 1760
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR