4-Bromo-2-nitroaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

B25468-14 B25468-22
AAB25468-14EA 57.6 USD
AAB25468-14 AAB25468-22
4-Bromo-2-nitroaniline 98%
4-Bromo-2-nitroaniline

Purity: 98% CAS No.: 875-51-4 Molecular Formula: C6H5BrN2O2 Formula Weight: 217.02 Melting Point: 111-114° MDL No.: MFCD00041312

Formula: C₆H₅BrN₂O₂
Boiling Pt: 309 °C (1013 hPa)
Melting Pt: 111…114 °C
Density: 1.81 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041312
CAS Number: 875-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR