5-Bromo-2-chlorophenol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25452-03
B25452-06
AAB25452-03EA
65.3
USD
AAB25452-03
AAB25452-06
5-Bromo-2-chlorophenol 98+%
5-Bromo-2-chlorophenol
Purity: 98+%
CAS No.: 183802-98-4
Molecular Formula: C6H4BrClO
Formula Weight: 207.46
Melting Point: 56-58°
MDL No.: MFCD00672940
Formula:
C₆H₄BrClO MW: 207.45 g/mol Melting Pt: 56…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00672940 CAS Number: 183802-98-4 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...